Order Entry
Netherlands
ContactUsLinkComponent
 
 
Tricyclo[6.4.0.0(2,7)]dodecane-1,8:2,7-tetracarboxylic dianhydride ≥98.0% (by GC)
 :  TCI
 
 
Tricyclo[6.4.0.0(2,7)]dodecane-1,8:2,7-tetracarboxylic dianhydride ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %

 

 

Frequently Bought Together