Tri-tert-butyl-1,4,7,10-tetraazacyclododecane-1,4,7,10-tetraacetate ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
Synonyms: DOTA(OtBu)₃, 2-(4,7,10-Tris(2-(tert-butoxy)-2-oxoethyl)-1,4,7,10-tetraazacyclododecan-1-yl)acetic acid, 1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic Acid Tri-tert-butyl Ester, 4,7,10-Tri-(t-butyloxycarbonylmethyl)-1,4,7,10-tetraazacyclododecan-1-yl-acetic acid hydrobromide salt, DOTA-tri(t-butyl ester)
CAS Number:137076-54-1
:
Formula:C₂₈H₅₂N₄O₈
MDL Number:MFCD02259697
:572.74 g/mol
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Gray to Red powder to crystal
- Purity (Chelometric Titration):min. 97.0 %
- Purity (Neutralization titration):min. 97.0 %
Specification Test Results
Frequently Bought Together