
2,2,2-Trifluoro-N-(4-methoxyphenyl)acetimidoyl chloride ≥98.0% (by GC, titration analysis)
Catalog # TCIAT2638-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Light orange to Yellow to Green clear liquid
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
About this item
:
:237.61 g/mol
Boiling Pt:98°C
Storage Temperature:Refrigerator
UN:3265
ADR:8,III
:GHS05
CAS Number:75999-66-5
Formula:C₉H₇ClF₃NO
MDL Number:MFCD09030149