(R)-(-)-2,3,7,7a-Tetrahydro-7a-methyl-1H-indene-1,5(6H)-dione ≥98.0% (by GC)
Catalog # TCIAT2636-1G
Leverancier: TCI
CAS-nummer:
Specificaties
- Pk:1 g
- Seq:3047
Specificaties
Specificatie testresultaten
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:57.0 to 61.0 deg-C
- Specific rotation [a]20/D:-284.0 to -315.0 deg (C=1, Toluene)
Specificatie testresultaten
Over dit artikel
:
:164.20 g/mol
CAS-nummer:17553-89-8
Formule:C₁₀H₁₂O₂
MDL-nummer:MFCD08235115