2,3,7,7a-Tetrahydro-7a-methyl-1H-indene-1,5(6H)-dione ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:19576-08-0
:
Formula:C₁₀H₁₂O₂
MDL Number:MFCD00135278
:164.20 g/mol
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:69.0 to 73.0 deg-C
Specification Test Results
Frequently Bought Together