Order Entry
Netherlands
ContactUsLinkComponent
Trimethoxy(p-tolyl)silane ≥97.0% (by GC)
Irritant
Trimethoxy(p-tolyl)silane ≥97.0% (by GC)
Catalog # TCIAT2560-5G
Supplier:  TCI
CAS Number:  
Irritant
Trimethoxy(p-tolyl)silane ≥97.0% (by GC)
Catalog # TCIAT2560-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :212.32 g/mol
Boiling Pt:143°C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:17873-01-7
Formula:C₁₀H₁₆O₃Si
MDL Number:MFCD00048020