Order Entry
Netherlands
ContactUsLinkComponent
(S)-(p-Toluenesulphinyl)ferrocene ≥97.0% (by HPLC, titration analysis)
(S)-(p-Toluenesulphinyl)ferrocene ≥97.0% (by HPLC, titration analysis)
Catalog # TCIAT2546-1G
Supplier:  TCI
CAS Number:  
(S)-(p-Toluenesulphinyl)ferrocene ≥97.0% (by HPLC, titration analysis)
Catalog # TCIAT2546-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Optical purity (LC):
    min. 97.0 ee%
  • Purity (Chelometric Titration):
    min. 97.0 %
  • Melting point:
    141.0 to 145.0 deg-C

Specification Test Results

About this item

 :
 :324.22 g/mol
CAS Number:164297-25-0
Formula:C₁₇H₁₆FeOS