(R)-(p-Toluenesulphinyl)ferrocene ≥98.0% (by HPLC, titration analysis)
: TCI
:130225-27-3
:
:C₁₇H₁₆FeOS
:324.22 g/mol
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Optical purity (LC):min. 98.0 ee%
- Purity (Chelometric Titration):min. 98.0 %
- Melting point:141.0 to 145.0 deg-C
Frequently Bought Together