Order Entry
Netherlands
ContactUsLinkComponent
N,N,5-Trimethylfurfurylamine ≥98.0% (by GC, titration analysis)
Flammable
Corrosive
N,N,5-Trimethylfurfurylamine ≥98.0% (by GC, titration analysis)
Catalog # TCIAT2508-5G
Supplier:  TCI
CAS Number:  
Flammable
Corrosive
N,N,5-Trimethylfurfurylamine ≥98.0% (by GC, titration analysis)
Catalog # TCIAT2508-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Amber to Dark green clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :139.20 g/mol
Boiling Pt:164°C
Flash Pt:49°C
UN:2733
ADR:3,III
 :GHS02
 :GHS05
CAS Number:14496-35-6
Formula:C₈H₁₃NO
MDL Number:MFCD00014113