Order Entry
Netherlands
ContactUsLinkComponent
1-(Trimethylsilyl)-2-naphthyl trifluoromethanesulphonate ≥96.0% (by GC)
Corrosive
1-(Trimethylsilyl)-2-naphthyl trifluoromethanesulphonate ≥96.0% (by GC)
Supplier:  TCI
Certificates
Corrosive
1-(Trimethylsilyl)-2-naphthyl trifluoromethanesulphonate ≥96.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 96.0 %

Specification Test Results

Frequently Bought Together