Order Entry
Netherlands
ContactUsLinkComponent
4,4',4''-Triaminotriphenylamine ≥98.0% (by HPLC, titration analysis)
Irritant
4,4',4''-Triaminotriphenylamine ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
Irritant
4,4',4''-Triaminotriphenylamine ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Gray to Brown powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

Related Information

Frequently Bought Together