

Tris(2-acryloyloxyethyl)isocyanurate ≥80.0% (by GC) stabilised
Supplier: TCI
Certificates
About this item
CAS Number:40220-08-4
:
Formula:C₁₈H₂₁N₃O₉
ADR:9,III
:Stabilised with phenothiazine
:423.38 g/mol
EINECS:254-843-6
UN:3077
:GHS07
:GHS09
Specification Test Results
- Appearance:White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
- Purity (GC):min. 80.0 %
Specification Test Results
Frequently Bought Together