Order Entry
Netherlands
ContactUsLinkComponent
4-(p-Tolyl)morpholine ≥97.0% (by GC)
Irritant
4-(p-Tolyl)morpholine ≥97.0% (by GC)
Catalog # TCIAT2215-5G
Supplier:  TCI
CAS Number:  
Irritant
4-(p-Tolyl)morpholine ≥97.0% (by GC)
Catalog # TCIAT2215-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Light orange to Yellow to Green powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Purity (Nonaqueous Titration):
    min. 95.0 %
  • Melting point:
    48.0 to 52.0 deg-C

Specification Test Results

About this item

 :
 :177.25 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:3077-16-5
Formula:C₁₁H₁₅NO
MDL Number:MFCD00222003