
N-(p-Tolyl)-1-naphthylamine ≥96.0% (by GC)
Catalog # TCIAT2201-500MG
Supplier: TCI
CAS Number:
Specifications
- Pk:500 mg
- Seq:2725
Specifications
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (GC):min. 96.0 %
- Melting point:76.0 to 79.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:233.31 g/mol
Boiling Pt:360°C
Melting Pt:78°C
:GHS07
CAS Number:634-43-5
Formula:C₁₇H₁₅N
MDL Number:MFCD00035709