
Tri-p-tolylsulphonium hexafluorophosphate ≥90.0% (by HPLC)
Catalog # TCIAT2041-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Purity (HPLC):min. 90.0 area%
- Purity (Ion exchange titration):min. 97.0 %
- Melting point:170.0 to 175.0 deg-C
Specification Test Results
About this item
:
:450.42 g/mol
:GHS07
CAS Number:146062-15-9
Formula:C₂₁H₂₁F₆PS
MDL Number:MFCD06797187