Order Entry
Netherlands
ContactUsLinkComponent
Tetraethylene glycol monobenzyl ether ≥95.0% (by GC)
Tetraethylene glycol monobenzyl ether ≥95.0% (by GC)
Supplier:  TCI
Certificates
Tetraethylene glycol monobenzyl ether ≥95.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Yellow to Green clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

Related Information

Frequently Bought Together