Methyl-3,4,5-tris(benzyloxy)benzoate ≥98.0% (by HPLC)
TCIAT1870-5G
: TCI
:
- VPE:5 g
- Seq:3173
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:100.0 to 104.0 deg-C
- Absorbance:min. 0.230 (10mg/L、CH3CN、263.0 to 266.0 nm)
- Absorbance2:min. 1.100 (10mg/L、CH3CN、216.0 to 219.0 nm)
:
:454.52 g/mol
:70424-94-1
:C₂₉H₂₆O₅
:MFCD03425302