Calcium-L-threonate ≥98.0% (by titrimetric analysis)
TCIAT1799-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:White to Almost white powder to crystal
- Purity (Chelometric Titration):min. 98.0 %
- Specific rotation [a]20/D:+14.0 to +16.0 deg (C=1, H2O)
:
:310.27 g/mol
:70753-61-6
:C₄H₈O₅
:MFCD00150824