Triphenylborane triphenylphosphine complex ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:3053-68-7
:
Formula:C₃₆H₃₀BP
MDL Number:MFCD01631317
:504.42 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 97.0 %
Specification Test Results
Frequently Bought Together