1,3,4,6-Tetra-O-acetyl-2-azido-2-deoxy-ɑ-D-galactopyranose
Catalog # TCIAT1731-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Melting point:120.0 to 127.0 deg-C
- Specific rotation [a]20/D:+98.0 to +102.0 deg (C=1, CHCl3)
- Elemental analysis (Nitrogen):11.00 to 11.40 %
Specification Test Results
About this item
:
:373.32 g/mol
Storage Temperature:Refrigerator
CAS Number:67817-30-5
Formula:C₁₄H₁₉N₃O₉
MDL Number:MFCD01076182