
5,10,15,20-Tetrakis(pentafluorophenyl)porphyrin ≥95.0% (by HPLC)
Catalog # TCIAT1730-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Gray to Dark purple to Black powder to crystal
- Purity (HPLC):min. 95.0 area%
Specification Test Results
About this item
:
:974.56 g/mol
:GHS07
CAS Number:25440-14-6
Formula:C₄₄H₁₀F₂₀N₄
MDL Number:MFCD00010032