
4-Acetylphenyl trifluoromethanesulphonate ≥98.0% (by GC)
Catalog # TCIAT1709-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
About this item
:
:268.21 g/mol
:GHS07
CAS Number:109613-00-5
Formula:C₉H₇F₃O₄S
MDL Number:MFCD00191706