Order Entry
Netherlands
ContactUsLinkComponent
o-Tolylmagnesium Bromide ∼17% ∼0.9 mol/L in THF
Flammable
Corrosive
Irritant
Health
o-Tolylmagnesium Bromide ∼17% ∼0.9 mol/L in THF
Catalog # TCIAT1698-100G
Supplier:  TCI
CAS Number:  
Flammable
Corrosive
Irritant
Health
o-Tolylmagnesium Bromide ∼17% ∼0.9 mol/L in THF
Catalog # TCIAT1698-100G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Concentration (sec Butanol method):
    17.0 to 20.0 w/w%

Specification Test Results

About this item

 :
 :195.34 g/mol
EINECS:213-250-2
UN:2924
ADR:3,II
 :GHS02
 :GHS05
 :GHS07
 :GHS08
CAS Number:932-31-0
Formula:C₇H₇BrMg
MDL Number:MFCD00010350