1,1,2,2-Tetrakis(4-methoxyphenyl)-1,2-ethanediol ≥96.0% (by HPLC)
TCIAT1651-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 96.0 area%
- Melting point:166.0 to 172.0 deg-C
- Solubility in Acetone:almost transparency
:
:486.56 g/mol
:19920-00-4
:C₃₀H₃₀O₆
:MFCD00185172