

Tris(trimethylsilyloxy)ethylene ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:69097-20-7
:
Formula:C₁₁H₂₈O₃Si₃
MDL Number:MFCD00011641
:292.60 g/mol
Flash Pt:39°C
Storage Temperature:Refrigerator
EINECS:273-864-1
UN:1993
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together