Order Entry
Netherlands
ContactUsLinkComponent
Triethyloxonium tetrafluoroborate 15% ∼1 mol/L in dichloromethane ethylating reagent
Corrosive
Irritant
Health
Environment
Triethyloxonium tetrafluoroborate 15% ∼1 mol/L in dichloromethane ethylating reagent
Catalog # TCIAT1606-100ML
Supplier:  TCI
CAS Number:  
undefined
Corrosive
Irritant
Health
Environment
Triethyloxonium tetrafluoroborate 15% ∼1 mol/L in dichloromethane ethylating reagent
Catalog # TCIAT1606-100ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Concentration (Nonaqueous Titration):
    15.0 to 16.5 w/w%

Specification Test Results

About this item

 :
 :189.99 g/mol
Merck Index:14,05796
Storage Temperature:Freezer
EINECS:206-705-1
UN:3265
ADR:8,II
 :GHS05
 :GHS07
 :GHS08
 :GHS09
CAS Number:368-39-8
Formula:C₆H₁₅BF₄O
MDL Number:MFCD00044423