

2,4,6-Triisopropylbenzenesulphonyl hydrazide ≥95.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:39085-59-1
:
Formula:C₁₅H₂₆N₂O₂S
MDL Number:MFCD00014750
Melting Pt:122°C
Storage Temperature:Freezer
EINECS:254-282-7
UN:1325
ADR:4.1,III
:298.45 g/mol
:GHS02
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Potassium bromate Method):min. 95.0 %
Specification Test Results
Frequently Bought Together