

2,4,6-Trimethoxyboroxin ≥96.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:102-24-9
:
Formula:C₃H₉B₃O₆
MDL Number:MFCD00005946
Flash Pt:9°C
EINECS:203-016-8
UN:2924
ADR:3,II
:173.53 g/mol
:GHS02
:GHS05
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (Neutralization titration):min. 96.0 %
Specification Test Results
Related Information
Frequently Bought Together