Order Entry
Netherlands
ContactUsLinkComponent
(Trimethylsilyl)methyl trifluoromethanesulphonate ≥98.0% (by GC) trimethylsilylmethylating reagent
Flammable
Corrosive
(Trimethylsilyl)methyl trifluoromethanesulphonate ≥98.0% (by GC) trimethylsilylmethylating reagent
Supplier:  TCI
Certificates
Flammable
Corrosive
(Trimethylsilyl)methyl trifluoromethanesulphonate ≥98.0% (by GC) trimethylsilylmethylating reagent
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 98.0 %
  • High boiling impurity of Silylation agent:
    No impurity higher than Isopropyl myristate (50 ppm) after 5 min + hold up time

Specification Test Results

Frequently Bought Together