
(3-Trimethylsilyl-2-propynyl)triphenylphosphonium bromide ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:42134-49-6
:
Formula:C₂₄H₂₆BrPSi
MDL Number:MFCD00012029
:453.43 g/mol
:GHS07
Specification Test Results
- Purity (HPLC):min. 96.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together