Order Entry
Netherlands
ContactUsLinkComponent
3,3,3',3'-Tetramethyl-5,5',6,6'-tetrapropoxy-1,1'-spirobiindane ≥98.0% (by HPLC)
3,3,3',3'-Tetramethyl-5,5',6,6'-tetrapropoxy-1,1'-spirobiindane ≥98.0% (by HPLC)
Catalog # TCIAT1477-25G
Supplier:  TCI
CAS Number:  
3,3,3',3'-Tetramethyl-5,5',6,6'-tetrapropoxy-1,1'-spirobiindane ≥98.0% (by HPLC)
Catalog # TCIAT1477-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Melting point:
    80.0 to 84.0 deg-C

Specification Test Results

About this item

 :
 :508.74 g/mol
CAS Number:89929-65-7
Formula:C₃₃H₄₈O₄
MDL Number:MFCD00191503