

N,N,N',N'-Tetramethyl-2-butene-1,4-diamine ≥97.0% (by GC, titration analysis)
Catalog # TCIAT1436-10ML
Supplier: TCI
CAS Number:
Specifications
- Pk:10 mL
- Seq:2275
Specifications
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
Specification Test Results
About this item
:
:142.25 g/mol
Flash Pt:50°C
EINECS:224-928-2
UN:2734
ADR:8,II
:GHS02
:GHS05
CAS Number:4559-79-9
Formula:C₈H₁₈N₂
MDL Number:MFCD00008336