Order Entry
Netherlands
ContactUsLinkComponent
3,3-Tetramethyleneglutarimide ≥98.0% (by HPLC, total nitrogen)
Toxic
Environment
3,3-Tetramethyleneglutarimide ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAT1412-10G
Supplier:  TCI
CAS Number:  
Toxic
Environment
3,3-Tetramethyleneglutarimide ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAT1412-10G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    153.0 to 156.0 deg-C

Specification Test Results

About this item

 :
ADR:6.1,III
 :167.21 g/mol
EINECS:427-770-4
UN:2811
 :GHS06
 :GHS09
CAS Number:1075-89-4
Formula:C₉H₁₃NO₂
MDL Number:MFCD00023871