
Tetrabutylammonium bis(1,3-dithiole-2-thione-4,5-dithiolato)nickel(III)complex ≥98.0% (by titrimetric analysis)
Catalog # TCIAT1272-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Purity (HPLC):min. 97.0 area%
- Purity (Chelometric Titration):min. 98.0 %
Specification Test Results
About this item
:
:693.83 g/mol
:GHS08
CAS Number:68401-88-7
Formula:C₂₂H₃₆NNiS₁₀
MDL Number:MFCD00059124