Order Entry
Netherlands
ContactUsLinkComponent
Triethylamine borane complex ≥90.0% (by titrimetric analysis)
 
 
 
Triethylamine borane complex ≥90.0% (by titrimetric analysis)
  TCIAT1180-25G
 :  TCI
 :  
 
 
 
Triethylamine borane complex ≥90.0% (by titrimetric analysis)
  TCIAT1180-25G
 :  TCI
 :  

 

  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (Iodometric Titration):
    min. 90.0 %

 

 

 :
 :115.03 g/mol
 :100°C
 :-7°C
 :–7°C
 :217-022-3
 :2924
 :3,II
 :GHS02
 :GHS05
 :GHS07
 :1722-26-5
 :C₆H₁₈BN
 :MFCD00012423