
Tetrahydroxy-1,4-benzoquinone hydrate ≥96.0% (by titrimetric analysis)
TCIAT1090-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:Dark green to Black powder to crystal
- Purity (Neutralization titration):min. 96.0 % (calcd.on dried substance)
- Drying loss:max. 18.0 %
:
:190.11 g/mol
:14,09249
:GHS07
:123334-16-7
:C₆H₆O₇
:MFCD00001597