
4,4',4''-Tris(benzoyloxy)trityl bromide ≥97.0% (by titrimetric analysis) hydroxyl protecting agent
Supplier: TCI
Certificates
About this item
CAS Number:86610-66-4
:
Formula:C₄₀H₂₇BrO₆
MDL Number:MFCD00010302
:683.55 g/mol
:GHS07
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (Argentmetric Titration):min. 97.0 %
- Melting point:192.0 to 198.0 deg-C
Specification Test Results
Frequently Bought Together