
1,1'-Thiobis(2-naphthol) ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAT1004-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:216.0 to 223.0 deg-C
- Solubility in N,N-DMF:almost transparency
Specification Test Results
About this item
:
:318.39 g/mol
EINECS:241-166-6
:GHS07
CAS Number:17096-15-0
Formula:C₂₀H₁₄O₂S
MDL Number:MFCD00046469