
Thujone (α and β mixture) ≥70% (by GC)
: TCI
:1125-12-8
:
:C₁₀H₁₆O
:MFCD00135597
:152.24 g/mol
:14,09392
:64°C
:214-405-7
:GHS07
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 70.0 % (total of isomer)
- Specitic rotation:-12.0 to -18.0 deg (neat)
Frequently Bought Together