
2,2',5,5'-Tetrachlorobenzidine ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:15721-02-5
:
Formula:C₁₂H₈Cl₄N₂
MDL Number:MFCD00043918
:322.01 g/mol
EINECS:239-815-3
:GHS07
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:137.0 to 141.0 deg-C
Specification Test Results
Frequently Bought Together