
Triethylene glycol monobutyl ether ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:143-22-6
:
Formula:C₁₀H₂₂O₄
MDL Number:MFCD00020606
:206.28 g/mol
Boiling Pt:278°C
Flash Pt:156°C
EINECS:205-592-6
:GHS05
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together