Triethylene glycol monododecyl ether ≥95.0% (by GC)
Catalog # TCIAT0768-5ML
Supplier: TCI
CAS Number:
Specifications
- Pk:5 mL
- Seq:2210
Specifications
Specification Test Results
- Appearance:White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
About this item
:
:318.50 g/mol
EINECS:221-280-2
CAS Number:3055-94-5
Formula:C₁₈H₃₈O₄
MDL Number:MFCD00042659