


Bis(4-aminophenyl)sulphide ≥98.0% (by HPLC, titration analysis)
TCIAT0632-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:White to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:108.0 to 111.0 deg-C
- Solubility in Methanol:almost transparency
:
:216.30 g/mol
:205-370-9
:3077
:9,III
:GHS07
:GHS08
:GHS09
:139-65-1
:C₁₂H₁₂N₂S
:MFCD00025342