endo-Tetrahydrodicyclopentadiene ≥97.0% (by GC)
Catalog # TCIAT0409-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:136.23 g/mol
Boiling Pt:192°C
EINECS:220-586-3
CAS Number:2825-83-4
Formula:C₁₀H₁₆
MDL Number:MFCD00074748