Dioctadecyl-3,3'-thiodipropionate ≥90.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:693-36-7
:
Formula:C₄₂H₈₂O₄S
MDL Number:MFCD00026684
:683.17 g/mol
EINECS:211-750-5
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC) (CAD):min. 90.0 area%
- Melting point:65.0 to 69.0 deg-C
- Solubility in Toluene:almost transparency
Specification Test Results
Frequently Bought Together