

Tetraphenylarsonium chloride hydrochloride hydrate ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAT0162-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 98.0 % (calcd.on anh.substance)
- Water:max. 10.0 %
Specification Test Results
About this item
:
:455.25 g/mol
UN:3465
ADR:6.1,III
:GHS06
:GHS09
CAS Number:123334-18-9
Formula:(C₆H₅)₄AsCl·HCl·xH₂O
MDL Number:MFCD00012628