


Tetraethylenepentamine (mixture of branched chain isomers and cyclic compounds)
Supplier: TCI
Certificates
About this item
CAS Number:112-57-2
:
Formula:C₈H₂₃N₅
MDL Number:MFCD00008168
:189.31 g/mol
Melting Pt:–40°C
Flash Pt:163°C
EINECS:203-986-2
UN:2320
ADR:8,III
:GHS05
:GHS07
:GHS09
Specification Test Results
- Appearance:Colorless to Yellow to Green clear liquid
- Purity (GC):many component mixture
Specification Test Results
Frequently Bought Together