Tetrabromofluorescein potassium salt ≥85.0% (by HPLC)
TCIAT0036-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:Orange to Brown to Dark red powder to crystal
- Purity (HPLC):min. 85.0 area%
:
:724.08 g/mol
:56897-54-2
:C₂₀H₈Br₄O₅
:MFCD00070510