Calcium-DL-tartrate tetrahydrate ≥98.0% (by titrimetric analysis)
Catalog # TCIAT0002-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Chelometric Titration):min. 98.0 % (calcd.on anh.substance)
- Solubility in HCl:almost transparency
- Water:26.3 to 28.5 %
Specification Test Results
About this item
:
:188.15 g/mol
Formula:C₄H₆O₆
MDL Number:MFCD00151484