
N-Succinimidyl tetradecanoate ≥95.0% (by HPLC, total nitrogen)
: TCI
:69888-86-4
:
:C₁₈H₃₁NO₄
:MFCD00522592
:325.45 g/mol
:GHS07
:Freezer
- Purity (HPLC):min. 95.0 area%
- Purity (with Total Nitrogen):min. 95.0 %
- Melting point:82.0 to 86.0 deg-C
Frequently Bought Together